Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308116.1 | NP_001295045.1 | 10 | 2117 | ||
NM_024947.3 | NP_079223.3 | 10 | 2102 | |||
XM_005247789.2 | XP_005247846.1 | 11 | 2180 | |||
XM_005247791.3 | XP_005247848.1 | 11 | 2180 | |||
XM_006713753.1 | XP_006713816.1 | 11 | 2149 | |||
XM_006713754.3 | XP_006713817.1 | 11 | 2271 | |||
XM_006713756.2 | XP_006713819.1 | 9 | 1922 | |||
XM_011513186.2 | XP_011511488.1 | 10 | 2081 | |||
XM_011513187.2 | XP_011511489.1 | 9 | 2033 | |||
XM_011513188.2 | XP_011511490.1 | 10 | 2021 | |||
XM_011513189.2 | XP_011511491.1 | 9 | 1973 | |||
XM_017007235.1 | XP_016862724.1 | 11 | 2168 | |||
XM_017007236.1 | XP_016862725.1 | 11 | 2156 | |||
XM_017007237.1 | XP_016862726.1 | 9 | 1973 | |||
XM_017007238.1 | XP_016862727.1 | 8 | 1874 | |||
XM_017007239.1 | XP_016862728.1 | 8 | 1721 | |||
GenBank mRNA | AF380154.1 | - | 10 | 2039 | ||
AF444193.1 | - | 9 | 2054 | |||
AJ320486.3 | - | 10 | 2096 | |||
AK304411.1 | - | 9 | 1943 | |||
BC131772.1 | - | 10 | 2081 | 40123356 | ||
EF560717.1 | - | 10 | 2077 |