Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCF2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112732.2 | NP_001106203.2 | 3 | 404 | ||
NM_001320815.1 | NP_001307744.1 | 3 | 436 | |||
NM_001320816.1 | NP_001307745.1 | 3 | 613 | |||
NM_001320817.1 | NP_001307746.1 | 3 | 436 | |||
NM_024979.4 | NP_079255.4 | 3 | 436 | |||
XM_011537482.1 | XP_011535784.1 | 4 | 393 | |||
XM_011537483.2 | XP_011535785.1 | 4 | 513 | |||
XM_011537484.1 | XP_011535786.1 | 3 | 840 | |||
XM_011537485.1 | XP_011535787.1 | 3 | 512 | |||
XM_011537486.2 | XP_011535788.2 | 3 | 528 | |||
XM_011537487.1 | XP_011535789.1 | 3 | 433 | |||
XM_011537488.1 | XP_011535790.1 | 3 | 272 | |||
XM_011537490.1 | XP_011535792.1 | 3 | 806 | |||
XM_017020493.1 | XP_016875982.1 | 3 | 528 | |||
XM_017020494.1 | XP_016875983.1 | 3 | 528 | |||
XM_017020495.1 | XP_016875984.1 | 3 | 528 | |||
XM_017020497.1 | XP_016875986.1 | 3 | 806 | |||
XM_017020499.1 | XP_016875988.1 | 3 | 528 | |||
GenBank mRNA | AB002360.1 | - | 3 | 381 | ||
AB116074.1 | - | 3 | 292 | |||
AK295047.1 | - | 3 | 433 | |||
AK295670.1 | - | 3 | 246 | |||
AK307867.1 | - | 3 | 584 | |||
AK315969.1 | - | 3 | 404 | |||
BC011853.2 | - | 3 | 396 | 4138009 | ||
BC020208.1 | - | 3 | 396 | 4121441 | ||
CR988747.1 | - | 3 | 404 | |||
DA114166.1 | - | 3 | 438 |