Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAIAP2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025045.5 | NP_079321.3 | 3 | 299 | ||
XM_005261751.4 | XP_005261808.1 | 4 | 300 | |||
XM_011530379.2 | XP_011528681.1 | 3 | 416 | |||
XM_011530381.2 | XP_011528683.1 | 3 | 193 | |||
XM_011530382.2 | XP_011528684.1 | 3 | 581 | |||
XM_011530383.2 | XP_011528685.1 | 3 | 581 | |||
XM_011530384.2 | XP_011528686.1 | 4 | 519 | |||
GenBank mRNA | AK026235.1 | - | 3 | 257 | ||
AY358164.1 | - | 3 | 299 |