Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UXS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253875.1 | NP_001240804.1 | 8, 9 | 733 | ||
NM_001253876.1 | NP_001240805.1 | 3, 4 | 870 | |||
NM_025076.4 | NP_079352.2 | 8, 9 | 718 | |||
NR_045607.1 | 7, 8 | 669 | ||||
XM_006712775.3 | XP_006712838.1 | 8, 9 | 695 | |||
XM_011511902.2 | XP_011510204.2 | 8, 9 | 712 | |||
XM_011511903.2 | XP_011510205.2 | 8, 9 | 695 | |||
XM_011511904.2 | XP_011510206.1 | 5, 6 | 1040 | |||
XM_017005008.1 | XP_016860497.1 | 8, 9 | 695 | |||
XM_017005009.1 | XP_016860498.1 | 8, 9 | 680 | |||
XM_017005010.1 | XP_016860499.1 | 8, 9 | 680 | |||
XM_017005011.1 | XP_016860500.1 | 8, 9 | 820 | |||
XM_017005012.1 | XP_016860501.1 | 8, 9 | 755 | |||
XM_017005013.1 | XP_016860502.1 | 6, 7 | 1278 | |||
XM_017005014.1 | XP_016860503.1 | 7, 8 | 496 | |||
GenBank mRNA | AK027244.1 | - | 3, 4 | 870 | ||
AK075120.1 | - | 8, 9 | 680 | |||
AK075170.1 | - | 8, 9 | 695 | |||
AK122696.1 | - | 7, 8 | 634 | |||
AK225592.1 | - | 3, 4 | 870 | |||
AK290668.1 | - | 8, 9 | 693 | |||
AK304872.1 | - | 2, 3 | 210 | |||
AY147934.1 | - | 8, 9 | 685 | |||
AY358541.1 | - | 8, 9 | 669 | |||
BC009819.2 | - | 8, 9 | 659 | 3843312 | ||
BI825770.1 | - | 8, 9 | 735 | |||
CF242897.1 | - | 8, 9 | 659 |