Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256599.1 | NP_001243528.1 | 4 | 891 | ||
NM_001256600.1 | NP_001243529.1 | 5 | 922 | |||
NM_001256601.1 | NP_001243530.1 | 4 | 722 | |||
NM_001282378.1 | NP_001269307.1 | 3 | 619 | |||
NM_001282379.1 | NP_001269308.1 | 4 | 881 | |||
NM_025058.4 | NP_079334.3 | 4 | 881 | |||
NR_046327.1 | 4 | 881 | ||||
NR_046329.1 | 5 | 940 | ||||
NR_104150.1 | 4 | 881 | ||||
XM_011510002.1 | XP_011508304.1 | 5 | 824 | |||
XM_017002393.1 | XP_016857882.1 | 5 | 995 | |||
GenBank mRNA | AK026882.1 | - | 4 | 792 | ||
AK096158.1 | - | 4 | 769 | |||
AK128139.1 | - | 5 | 940 | |||
AK225075.1 | - | 4 | 792 | |||
AK296300.1 | - | 5 | 918 | |||
AK297318.1 | - | 3 | 527 | |||
AK304542.1 | - | 4 | 725 | |||
AK308075.1 | - | 4 | 686 | |||
AK309699.1 | - | 4 | 722 | |||
AK310429.1 | - | 5 | 1027 | |||
AY251386.1 | - | 4 | 701 | |||
BC069416.1 | - | 4 | 777 | 7262300 | ||
BC069568.1 | - | 4 | 777 | 7262312 | ||
BC126372.1 | - | 4 | 798 | 8992088 | ||
BC143828.1 | - | 4 | 798 | |||
CN259033.1 | - | 3 | 571 | |||
DB262381.1 | - | 2 | 371 |