Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005334.2 | NP_005325.2 | 3, 4 | 1039 | ||
XM_006724815.2 | XP_006724878.1 | 3, 4 | 1504 | |||
XM_006724816.2 | XP_006724879.1 | 3, 4 | 1504 | |||
XM_011531147.2 | XP_011529449.1 | 3, 4 | 1504 | |||
XM_011531148.2 | XP_011529450.1 | 3, 4 | 1504 | |||
XM_017029471.1 | XP_016884960.1 | 3, 4 | 1504 | |||
GenBank mRNA | BC019887.1 | - | 3, 4 | 574 | ||
BC030560.1 | - | 3, 4 | 660 | |||
BC063435.1 | - | 3, 4 | 1050 | |||
DQ655939.2 | - | 2, 3 | 349 | |||
L20010.1 | - | 3, 4 | 855 |