Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIKE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102396.1 | NP_001095866.1 | 2 | 286 | ||
NM_025073.2 | NP_079349.2 | 2 | 274 | |||
NR_049741.1 | 2 | 336 | ||||
GenBank mRNA | AK024821.1 | - | 2 | 264 | ||
AK126316.1 | - | 2 | 284 | |||
AK225734.1 | - | 2 | 265 | |||
BC005934.1 | - | 2 | 244 | 4249034 | ||
CR749219.1 | - | 2 | 277 | |||
CR936771.1 | - | 2 | 277 | |||
DA950119.1 | - | 1 | 511 | |||
DB459255.1 | - | 2 | 104 |