Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCLS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292041.1 | NP_001278970.1 | 4 | 326 | ||
NM_005335.5 | NP_005326.2 | 4 | 326 | |||
GenBank mRNA | AK225633.1 | - | 3 | 199 | ||
AK298663.1 | - | 4 | 305 | |||
AK298696.1 | - | 4 | 276 | |||
AK300301.1 | - | 4 | 276 | |||
AK312750.1 | - | 4 | 289 | |||
BC016758.2 | - | 4 | 246 | 2966303 | ||
BT006824.1 | - | 3 | 197 | |||
CR456794.1 | - | 3 | 197 | |||
DC407066.1 | - | 4 | 322 | |||
DC427610.1 | - | 4 | 327 | |||
X16663.1 | - | 4 | 239 |