Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099222.1 | NP_001092692.1 | 9, 10 | 905 | ||
NM_001099223.1 | NP_001092693.1 | 9, 10 | 861 | |||
NM_001099224.1 | NP_001092694.1 | 9, 10 | 897 | |||
NM_025103.2 | NP_079379.2 | 9, 10 | 897 | |||
XM_011518035.2 | XP_011516337.1 | 9, 10 | 897 | |||
XM_011518036.2 | XP_011516338.1 | 2, 3 | 607 | |||
XM_017015163.1 | XP_016870652.1 | 2, 3 | 412 | |||
XM_017015164.1 | XP_016870653.1 | 2, 3 | 1853 | |||
GenBank mRNA | AK023707.1 | - | 9, 10 | 858 | ||
AK225570.1 | - | 9, 10 | 858 | |||
AY040325.1 | - | 9, 10 | 826 | |||
BC023634.2 | - | 9, 10 | 842 | |||
BC056875.1 | - | 7, 8 | 605 | |||
BC107742.1 | - | 9, 10 | 795 | 6614193 | ||
BC113524.1 | - | 8, 9 | 670 | |||
BC113526.1 | - | 8, 9 | 671 |