Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NYNRIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025081.2 | NP_079357.2 | 9 | 4951 | ||
XM_011537016.2 | XP_011535318.1 | 4 | 3226 | |||
GenBank mRNA | AB037726.2 | - | 7 | 4396 | ||
AK021873.1 | - | 3 | 736 |