Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGGHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025092.4 | NP_079368.3 | 14 | 2234 | ||
XM_011520383.2 | XP_011518685.1 | 17 | 3160 | |||
XM_011520384.2 | XP_011518686.1 | 16 | 3217 | |||
XM_017018355.1 | XP_016873844.1 | 15 | 2612 | |||
GenBank mRNA | AK026288.1 | - | 7 | 1019 | ||
AK074155.1 | - | 11 | 3396 | |||
AK090428.1 | - | 8 | 1851 | |||
AK296137.1 | - | 10 | 1113 | |||
AL832932.1 | - | 7 | 892 | |||
BC048336.1 | - | 5 | 781 | |||
BC066641.1 | - | 13 | 1941 | |||
BC109257.2 | - | 11 | 1633 | 40024867 | ||
BC109258.2 | - | 11 | 1633 | 40024874 |