Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033580.2 | NP_001028752.1 | 4 | 1093 | ||
NM_001163735.1 | NP_001157207.1 | 4 | 1093 | |||
NM_025109.5 | NP_079385.2 | 4 | 1093 | |||
XM_011525286.1 | XP_011523588.1 | 4 | 549 | |||
XM_011525287.1 | XP_011523589.1 | 4 | 549 | |||
XM_011525290.2 | XP_011523592.1 | 4 | 549 | |||
XM_017025157.1 | XP_016880646.1 | 4 | 523 | |||
XM_017025158.1 | XP_016880647.1 | 4 | 549 | |||
XM_017025159.1 | XP_016880648.1 | 4 | 549 | |||
XM_017025160.1 | XP_016880649.1 | 4 | 549 | |||
XM_017025161.1 | XP_016880650.1 | 4 | 504 | |||
GenBank mRNA | AB209035.1 | - | 4 | 1051 | ||
AK026518.1 | - | 4 | 504 | |||
AK293903.1 | - | 4 | 1093 | |||
AK304073.1 | - | 4 | 444 | |||
BC008900.2 | - | 4 | 484 | 3629870 | ||
BM921100.1 | - | 546 |