Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102401.2 | NP_001095871.1 | 2 | 295 | ||
NM_001265581.1 | NP_001252510.1 | 2 | 417 | |||
NM_025115.3 | NP_079391.2 | 1 | 664 | |||
XM_011544665.2 | XP_011542967.1 | 2 | 711 | |||
GenBank mRNA | AK026916.1 | - | 1 | 517 | ||
AL530920.3 | - | 1 | 193 | |||
BC007387.2 | - | 2 | 220 | 3830500 | ||
BC066935.1 | - | 2 | 248 | 4839169 | ||
BX648410.1 | - | 2 | 331 | |||
DB047280.1 | - | 2 | 308 | |||
DB112682.1 | - | 1 | 561 | |||
DB113350.1 | - | 1 | 472 |