Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025126.3 | NP_079402.2 | 2 | 307 | ||
NM_194271.2 | NP_919247.1 | 3 | 338 | |||
GenBank mRNA | AB084914.1 | - | 1 | 180 | ||
AF306709.1 | - | 2 | 349 | |||
AK025439.1 | - | 2 | 272 | |||
AK225285.1 | - | 2 | 272 | |||
AK304366.1 | - | 3 | 327 | |||
AL557811.3 | - | 3 | 348 | |||
AY098934.1 | - | 1 | 177 | |||
BC007826.2 | - | 2 | 232 | 3347885 | ||
BT007283.1 | - | 1 | 177 | |||
CR457342.1 | - | 1 | 177 | |||
DA887995.1 | - | 3 | 338 | |||
DB242808.1 | - | 2 | 308 |