Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304957.1 | NP_001291886.1 | 4, 5 | 737 | ||
NM_001304958.1 | NP_001291887.1 | 5, 6 | 844 | |||
NM_001304959.1 | NP_001291888.1 | 4, 5 | 486 | |||
NM_001304960.1 | NP_001291889.1 | 6, 7 | 655 | |||
NM_001304961.1 | NP_001291890.1 | 3, 4 | 654 | |||
NM_025140.2 | NP_079416.1 | 4, 5 | 747 | |||
XM_005253624.2 | XP_005253681.1 | 4, 5 | 491 | |||
XM_017019984.1 | XP_016875473.1 | 4, 5 | 1305 | |||
GenBank mRNA | AB015292.1 | - | 3, 4 | 267 | ||
AK026124.1 | - | 4, 5 | 378 | |||
AK054680.1 | - | 2, 3 | 2014 | |||
AK125866.1 | - | 2, 3 | 2265 | |||
AK222661.1 | - | 4, 5 | 335 | |||
AW953473.1 | - | 6, 8 | 626 | |||
BC017186.2 | - | 4, 5 | 370 | 3051346 | ||
BX394686.2 | - | 3, 4 | 403 | |||
DA263704.1 | - | 4, 5 | 486 | |||
DA370549.1 | - | 5, 6 | 442 |