Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP2K1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002755.3 | NP_002746.1 | 2 | 636 | ||
XM_011521783.2 | XP_011520085.1 | 2 | 293 | |||
XM_017022411.1 | XP_016877900.1 | 2 | 587 | |||
XM_017022412.1 | XP_016877901.1 | 2 | 173 | |||
GenBank mRNA | AK291500.1 | - | 2 | 597 | ||
AK294322.1 | - | 2 | 147 | |||
AK312356.1 | - | 2 | 543 | |||
BC137459.1 | - | 2 | 224 | 9021459 | ||
BC139729.1 | - | 2 | 220 | 40053655 | ||
L05624.1 | - | 2 | 194 | |||
L11284.1 | - | 2 | 233 |