Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145635.1 | NP_001139107.1 | 1 | 715 | ||
NM_025188.3 | NP_079464.2 | 1 | 715 | |||
GenBank mRNA | AK023243.1 | - | 1 | 654 | ||
AK222901.1 | - | 1 | 659 | |||
AY669488.1 | - | 1 | 715 | |||
BC034943.2 | - | 1 | 688 | 4826874 |