Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142319.1 | NP_001135791.1 | 2 | 467 | ||
NM_025203.2 | NP_079479.1 | 2 | 467 | |||
XM_017005029.1 | XP_016860518.1 | 2 | 462 | |||
GenBank mRNA | AK025598.1 | - | 2 | 394 | ||
BC035698.2 | - | 2 | 402 | 5547301 | ||
DB292134.1 | - | 2 | 469 |