Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184891.1 | NP_001171820.1 | 3 | 349 | ||
NM_001184892.1 | NP_001171821.1 | 3 | 988 | |||
NM_025207.4 | NP_079483.3 | 2 | 793 | |||
NM_201398.2 | NP_958800.1 | 3 | 349 | |||
GenBank mRNA | AF218022.1 | - | 3 | 915 | ||
AF481877.1 | - | 2 | 101 | |||
AF520568.1 | - | 2 | 714 | |||
AK025979.1 | - | 3 | 277 | |||
AK090542.1 | - | 2 | 735 | |||
AK308243.1 | - | 3 | 283 | |||
BC011378.1 | - | 3 | 278 | 3855223 | ||
BC014012.2 | - | 1 | 68 | |||
BC021096.1 | - | 3 | 267 | 4891331 | ||
BC032323.1 | - | 3 | 284 | 5242727 | ||
BI919061.1 | - | 3 | 331 | |||
DC371856.1 | - | 3 | 350 | |||
DQ458779.1 | - | 2 | 392 |