Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025222.3 | NP_079498.2 | 9 | 2727 | ||
XM_011534136.2 | XP_011532438.1 | 8 | 2323 | |||
GenBank mRNA | AF132207.1 | - | 1 | 768 | ||
AF173977.1 | - | 1 | 504 | |||
AK074290.1 | - | 5 | 1943 | |||
AK098435.1 | - | 7 | 2173 | |||
AK123860.1 | - | 7 | 3306 | |||
AK130177.1 | - | 1 | 518 | |||
BC011760.2 | - | 1 | 787 | |||
BC018941.1 | - | 5 | 1930 | |||
BX648628.1 | - | 9 | 2469 |