Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REEP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316964.1 | NP_001303893.1 | 2 | 552 | ||
NM_001316965.1 | NP_001303894.1 | 2 | 552 | |||
NM_025232.3 | NP_079508.2 | 2 | 552 | |||
GenBank mRNA | AF218014.1 | - | 2 | 330 | ||
AK025899.1 | - | 2 | 482 | |||
AK025930.1 | - | 2 | 185 | |||
AK172846.1 | - | 2 | 487 | |||
AK302352.1 | - | 2 | 369 | |||
AY562242.1 | - | 2 | 83 | |||
BC013048.1 | - | 2 | 536 | 3863838 | ||
BC050622.1 | - | 2 | 454 | 6504298 | ||
BG695963.1 | - | 2 | 141 | |||
BP351388.1 | - | 2 | 554 | |||
CR457353.1 | - | 2 | 83 |