Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNKS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025235.3 | NP_079511.1 | 2 | 708 | ||
XM_005270185.4 | XP_005270242.2 | 2 | 739 | |||
XM_011540213.1 | XP_011538515.1 | 2 | 739 | |||
XM_017016696.1 | XP_016872185.1 | 2 | 676 | |||
XM_017016697.1 | XP_016872186.1 | 2 | 676 | |||
XM_017016698.1 | XP_016872187.1 | 2 | 387 | |||
XM_017016699.1 | XP_016872188.1 | 2 | 676 | |||
XM_017016700.1 | XP_016872189.1 | 2 | 289 | |||
XM_017016701.1 | XP_016872190.1 | 2 | 739 | |||
GenBank mRNA | AF264912.1 | - | 2 | 411 | ||
AF305081.1 | - | 2 | 713 | |||
AF309033.1 | - | 2 | 662 | |||
AF329696.1 | - | 2 | 550 | |||
AF342982.1 | - | 2 | 411 | |||
AF438201.1 | - | 2 | 790 | |||
AK314612.1 | - | 2 | 676 |