Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD1LG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025239.3 | NP_079515.2 | 3 | 362 | ||
XM_005251600.3 | XP_005251657.1 | 3 | 469 | |||
GenBank mRNA | AF329193.1 | - | 3 | 347 | ||
AF344424.1 | - | 3 | 362 | |||
AY254343.1 | - | 2 | 89 | |||
AY271901.1 | - | 2 | 89 | |||
AY271902.1 | - | 2 | 89 | |||
BC074766.2 | - | 3 | 123 | 30915184 | ||
BC113678.1 | - | 3 | 125 | 8322730 | ||
BC113680.1 | - | 3 | 125 | 8322732 | ||
BC143920.1 | - | 3 | 125 | |||
DQ336699.1 | - | 2 | 56 | |||
EF444806.1 | - | 3 | 209 |