Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024736.1 | NP_001019907.1 | 4 | 839 | ||
NM_025240.2 | NP_079516.1 | 4 | 839 | |||
XM_005254700.4 | XP_005254757.1 | 4 | 957 | |||
XM_011522095.2 | XP_011520397.1 | 4 | 828 | |||
XM_011522096.2 | XP_011520398.1 | 4 | 768 | |||
XM_017022638.1 | XP_016878127.1 | 4 | 757 | |||
GenBank mRNA | AF302102.1 | - | 3 | 605 | ||
AJ583695.1 | - | 3 | 648 | |||
AK074849.1 | - | 4 | 729 | |||
AK074997.1 | - | 2 | 335 | |||
AK075549.1 | - | 4 | 767 | |||
AK291721.1 | - | 4 | 676 | |||
AK292711.1 | - | 3 | 600 | |||
AK296358.1 | - | 4 | 557 | |||
AL360136.1 | - | 1 | 167 | |||
AY358343.1 | - | 4 | 756 | |||
BC062581.1 | - | 4 | 839 | 5201491 | ||
BQ278722.1 | - | 422 |