Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSEN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145392.1 | NP_001138864.1 | 2 | 230 | ||
NM_001145393.2 | NP_001138865.1 | 2 | 198 | |||
NM_001145394.1 | NP_001138866.1 | 2 | 198 | |||
NM_001321277.1 | NP_001308206.1 | 2 | 198 | |||
NM_001321278.1 | NP_001308207.1 | 2 | 198 | |||
NM_001321279.1 | NP_001308208.1 | 2 | 464 | |||
NM_025265.3 | NP_079541.1 | 2 | 482 | |||
NR_135607.1 | 2 | 198 | ||||
XM_017007292.1 | XP_016862781.1 | 2 | 150 | |||
XM_017007293.1 | XP_016862782.1 | 2 | 482 | |||
XM_017007294.1 | XP_016862783.1 | 2 | 216 | |||
XM_017007295.1 | XP_016862784.1 | 2 | 412 | |||
XM_017007296.1 | XP_016862785.1 | 2 | 778 | |||
XM_017007297.1 | XP_016862786.1 | 2 | 482 | |||
GenBank mRNA | AK027821.1 | - | 2 | 482 | ||
AK074794.1 | - | 2 | 129 | |||
AK225875.1 | - | 2 | 187 | |||
AK291183.1 | - | 2 | 267 | |||
AK295508.1 | - | 2 | 438 | |||
AK300449.1 | - | 2 | 447 | |||
BC004178.2 | - | 2 | 215 | 2959536 | ||
BC004211.2 | - | 2 | 215 | 2959536 | ||
BC019582.1 | - | 2 | 464 | 3872331 | ||
DA063377.1 | - | 2 | 484 | |||
DA614108.1 | - | 2 | 199 |