Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGES3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142654.1 | NP_001136126.1 | 3, 4 | 645 | ||
NM_001261430.1 | NP_001248359.1 | 4, 5 | 759 | |||
GenBank mRNA | AK056965.1 | - | 3, 4 | 201 | ||
AK093402.1 | - | 2, 3 | 1069 | |||
BG776211.1 | - | 4, 5 | 306 |
Gene Symbol: PTGES3L-AARSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136042.2 | NP_001129514.2 | 4, 5 | 759 | ||
NM_025267.3 | NP_079543.1 | 4, 5 | 375 | |||
GenBank mRNA | AF039235.1 | - | 4, 5 | 464 | ||
AK094084.1 | - | 4, 5 | 313 | |||
AK295447.1 | - | 4, 5 | 759 | |||
BC004172.2 | - | 4, 5 | 375 | 2823004 | ||
BC019324.1 | - | 4, 5 | 375 | 2823004 |