Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK026916.1 | - | 1 | 566 | ||
BC007387.2 | - | 2 | 269 | 3830500 | ||
BX648410.1 | - | 2 | 380 | |||
DB047280.1 | - | 2 | 357 | |||
DB113350.1 | - | 1 | 521 |