Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190274.1 | NP_001177203.1 | 4 | 603 | ||
NM_025133.4 | NP_079409.3 | 4 | 392 | |||
XM_005264572.4 | XP_005264629.1 | 4 | 597 | |||
XM_005264573.4 | XP_005264630.1 | 4 | 550 | |||
XM_017005015.1 | XP_016860504.1 | 4 | 550 | |||
XM_017005016.1 | XP_016860505.1 | 4 | 416 | |||
XM_017005017.1 | XP_016860506.1 | 4 | 416 | |||
GenBank mRNA | AF174599.1 | - | 4 | 485 | ||
AF176706.1 | - | 4 | 473 | |||
AF351618.1 | - | 4 | 405 | |||
AK223592.1 | - | 4 | 487 | |||
AK292877.1 | - | 4 | 442 | |||
AK297444.1 | - | 4 | 577 | |||
AK304140.1 | - | 4 | 307 | |||
AK315108.1 | - | 4 | 392 | |||
AY827075.1 | - | 4 | 530 | |||
BC043258.1 | - | 4 | 494 | |||
BC130445.1 | - | 4 | 328 | 40146476 | ||
BC136480.1 | - | 4 | 328 | 9020469 | ||
DN994547.1 | - | 4 | 611 |