Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304957.1 | NP_001291886.1 | 5 | 886 | ||
NM_001304958.1 | NP_001291887.1 | 6 | 993 | |||
NM_001304959.1 | NP_001291888.1 | 5 | 635 | |||
NM_001304960.1 | NP_001291889.1 | 7 | 804 | |||
NM_001304961.1 | NP_001291890.1 | 4 | 803 | |||
NM_025140.2 | NP_079416.1 | 5 | 896 | |||
XM_005253624.2 | XP_005253681.1 | 5 | 640 | |||
XM_017019984.1 | XP_016875473.1 | 5 | 1454 | |||
GenBank mRNA | AB015292.1 | - | 4 | 416 | ||
AK026124.1 | - | 5 | 527 | |||
AK054680.1 | - | 3 | 2163 | |||
AK124358.1 | - | 1 | 1985 | |||
AK125866.1 | - | 3 | 2414 | |||
AK222661.1 | - | 5 | 484 | |||
BC017186.2 | - | 5 | 519 | 3051346 | ||
BX394686.2 | - | 4 | 552 |