Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAA50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308445.1 | NP_001295374.1 | 2 | 478 | ||
NM_025146.3 | NP_079422.1 | 2 | 481 | |||
GenBank mRNA | AK023090.1 | - | 2 | 396 | ||
AK023256.1 | - | 2 | 431 | |||
AK296456.1 | - | 2 | 176 | |||
AK307184.1 | - | 3 | 325 | |||
AK315876.1 | - | 2 | 283 | |||
AL601126.1 | - | 2 | 376 | |||
BC012731.2 | - | 2 | 420 | 3619903 | ||
DA854531.1 | - | 2 | 478 |