Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272004.1 | NP_001258933.1 | 1, 2 | 458 | ||
NM_025209.3 | NP_079485.1 | 1, 2 | 458 | |||
GenBank mRNA | AF277374.1 | - | 1, 2 | 425 | ||
AK309822.1 | - | 1, 2 | 327 | |||
BC036529.1 | - | 1, 2 | 459 | 4828057 | ||
BX476993.1 | - | 1, 2 | 449 | |||
CR749375.1 | - | 2, 3 | 152 | |||
DA333251.1 | - | 1, 2 | 442 |