Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006084.4 | NP_006075.3 | 2, 3 | 315 | ||
GenBank mRNA | AK295467.1 | - | 2, 3 | 208 | ||
AK308347.1 | - | 2, 3 | 238 | |||
BC035716.2 | - | 1, 2 | 191 | 5588434 | ||
BI919023.1 | - | 2, 3 | 502 | |||
M87503.1 | - | 2, 3 | 222 |