Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY6G5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025262.3 | NP_079538.3 | 3 | 338 | ||
GenBank mRNA | AJ315540.1 | - | 4 | 442 | ||
AJ315541.1 | - | 4 | 562 | |||
AJ315542.1 | - | 3 | 329 | |||
AJ315543.1 | - | 3 | 423 | |||
AJ315544.1 | - | 2 | 1313 |