Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0226L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286761.1 | NP_001273690.1 | 4, 5 | 1209 | ||
NM_001286762.1 | NP_001273691.1 | 3, 4 | 1115 | |||
NM_001286763.1 | NP_001273692.1 | 4, 5 | 844 | |||
NM_001286764.1 | NP_001273693.1 | 3, 4 | 728 | |||
NM_001286765.1 | NP_001273694.1 | 3, 4 | 574 | |||
NM_001286766.1 | NP_001273695.1 | 2, 3 | 458 | |||
NM_025113.3 | NP_079389.2 | 4, 5 | 1231 | |||
GenBank mRNA | AK025215.1 | - | 3, 4 | 864 | ||
AK093073.1 | - | 4, 5 | 1179 | |||
AK125950.1 | - | 4, 5 | 1216 | |||
AK293034.1 | - | 3, 4 | 480 | |||
AK300183.1 | - | 3, 4 | 278 | |||
AK307854.1 | - | 2, 3 | 201 | |||
AK310264.1 | - | 5, 6 | 1218 | |||
BC032311.1 | - | 3, 4 | 796 | |||
BC043488.1 | - | 4, 5 | 577 | 5205680 | ||
BX641126.1 | - | 2, 3 | 265 | |||
DC330997.1 | - | 2, 3 | 459 |