Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271895.1 | NP_001258824.1 | 16 | 2199 | ||
NM_001271896.1 | NP_001258825.1 | 14 | 2055 | |||
NM_025150.4 | NP_079426.2 | 18 | 2445 | |||
NR_073513.1 | 14 | 1844 | ||||
NR_073514.1 | 17 | 2320 | ||||
XM_006711555.1 | XP_006711618.1 | 17 | 2306 | |||
XM_006711556.1 | XP_006711619.1 | 17 | 2261 | |||
XM_017002394.1 | XP_016857883.1 | 16 | 2159 | |||
XM_017002395.1 | XP_016857884.1 | 15 | 2060 | |||
GenBank mRNA | AL512732.1 | - | 7 | 909 | ||
AY450558.1 | - | 14 | 1774 | |||
BC000541.2 | - | 18 | 2392 | 3163506 | ||
BC007824.2 | - | 18 | 2333 | |||
BC009997.2 | - | 18 | 2365 | 4128881 | ||
DB551795.1 | - | 1 | 127 |