Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000111.2 | NP_000102.1 | 18 | 2253 | ||
XM_011515867.2 | XP_011514169.1 | 18 | 2234 | |||
GenBank mRNA | AK300651.1 | - | 12 | 1281 | ||
BC025671.1 | - | 18 | 2227 | 5202612 | ||
BX640837.1 | - | 17 | 1962 | |||
L02785.1 | - | 18 | 2226 |