Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025165.2 | NP_079441.1 | 10 | 1473 | ||
GenBank mRNA | AF276512.1 | - | 10 | 1070 | ||
AK026290.1 | - | 10 | 1155 | |||
AK057528.1 | - | 8 | 1158 | |||
AK126384.1 | - | 10 | 1712 | |||
BC006548.2 | - | 10 | 1124 | 2821972 | ||
BC019293.1 | - | 10 | 1124 | 2821972 | ||
CR457358.1 | - | 10 | 1070 |