Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NINL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318226.1 | NP_001305155.1 | 23 | 3305 | ||
NM_025176.5 | NP_079452.3 | 24 | 4352 | |||
XM_011529186.2 | XP_011527488.1 | 25 | 5191 | |||
XM_011529187.2 | XP_011527489.1 | 25 | 5176 | |||
XM_011529188.1 | XP_011527490.1 | 25 | 4568 | |||
XM_011529189.1 | XP_011527491.1 | 25 | 4576 | |||
XM_011529190.1 | XP_011527492.1 | 23 | 4290 | |||
XM_011529191.1 | XP_011527493.1 | 21 | 4083 | |||
XM_011529192.1 | XP_011527494.1 | 18 | 5003 | |||
XM_011529193.1 | XP_011527495.1 | 22 | 3243 | |||
XM_011529194.2 | XP_011527496.1 | 16 | 4272 | |||
XM_011529195.2 | XP_011527497.1 | 15 | 3097 | |||
XM_011529197.2 | XP_011527499.1 | 17 | 3466 | |||
XM_011529198.1 | XP_011527500.1 | 11 | 2638 | |||
XM_017027732.1 | XP_016883221.1 | 26 | 6510 | |||
XM_017027733.1 | XP_016883222.1 | 26 | 4622 | |||
XM_017027734.1 | XP_016883223.1 | 18 | 3456 | |||
XM_017027735.1 | XP_016883224.1 | 15 | 3102 | |||
GenBank mRNA | AB023197.1 | - | 24 | 4338 | ||
AK021854.1 | - | 3 | 1014 | |||
BC036380.2 | - | 23 | 4284 | 5268454 | ||
EU718622.1 | - | 23 | 3291 |