Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35F5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025181.3 | NP_079457.2 | 16 | 2265 | ||
NR_104470.1 | 17 | 2534 | ||||
XM_005263799.3 | XP_005263856.1 | 16 | 2650 | |||
XM_011511921.2 | XP_011510223.1 | 17 | 2101 | |||
XM_011511922.2 | XP_011510224.1 | 17 | 2318 | |||
XM_011511923.2 | XP_011510225.1 | 16 | 2003 | |||
XM_017005028.1 | XP_016860517.1 | 15 | 1964 | |||
GenBank mRNA | AK024666.1 | - | 11 | 1393 | ||
AY358620.1 | - | 16 | 2008 | |||
BC018537.1 | - | 16 | 2275 | 4340152 |