Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTERF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033050.2 | NP_001028222.1 | 3 | 752 | ||
NM_025198.4 | NP_079474.2 | 3 | 742 | |||
XM_011538767.2 | XP_011537069.1 | 3 | 1835 | |||
XM_017019985.1 | XP_016875474.1 | 4 | 844 | |||
XM_017019986.1 | XP_016875475.1 | 4 | 834 | |||
XM_017019987.1 | XP_016875476.1 | 2 | 640 | |||
GenBank mRNA | AK024124.1 | - | 3 | 737 | ||
AK222558.1 | - | 3 | 711 | |||
AL832827.1 | - | 2 | 650 | |||
AY008301.1 | - | 3 | 704 | |||
BC025984.1 | - | 3 | 726 | 4106188 | ||
BC036066.1 | - | 3 | 2375 | 5313686 |