Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282447.2 | NP_001269376.1 | 22 | 3765 | ||
NM_025220.4 | NP_079496.1 | 22 | 3768 | |||
NM_153202.3 | NP_694882.1 | 21 | 3690 | |||
XM_005260843.1 | XP_005260900.1 | 22 | 3225 | |||
XM_006723639.1 | XP_006723702.1 | 22 | 3222 | |||
XM_006723640.1 | XP_006723703.1 | 22 | 3216 | |||
XM_011529366.1 | XP_011527668.1 | 22 | 3124 | |||
XM_011529367.1 | XP_011527669.1 | 22 | 3085 | |||
XM_011529368.1 | XP_011527670.1 | 21 | 3147 | |||
XM_011529373.1 | XP_011527675.1 | 21 | 3199 | |||
GenBank mRNA | AF466287.1 | - | 22 | 3084 | ||
AK092627.1 | - | 8 | 1962 | |||
AK094070.1 | - | 2 | 2012 | |||
AK095953.1 | - | 14 | 2870 | |||
AK123015.1 | - | 18 | 3789 | |||
AK126487.1 | - | 16 | 4766 | |||
AK126498.1 | - | 10 | 4525 | |||
AK226148.1 | - | 12 | 4829 | |||
AL117415.1 | - | 11 | 2643 | |||
AY358314.1 | - | 22 | 3196 | |||
BX103461.1 | - | 1 | 380 |