Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSEN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145392.1 | NP_001138864.1 | 6 | 991 | ||
NM_001145394.1 | NP_001138866.1 | 7 | 782 | |||
NM_001321277.1 | NP_001308206.1 | 6 | 959 | |||
NM_001321278.1 | NP_001308207.1 | 6 | 959 | |||
NM_025265.3 | NP_079541.1 | 6 | 1243 | |||
NR_135607.1 | 6 | 959 | ||||
XM_011534139.2 | XP_011532441.1 | 4 | 1142 | |||
XM_017007292.1 | XP_016862781.1 | 7 | 968 | |||
XM_017007293.1 | XP_016862782.1 | 7 | 1300 | |||
XM_017007294.1 | XP_016862783.1 | 7 | 1034 | |||
XM_017007295.1 | XP_016862784.1 | 7 | 1230 | |||
XM_017007296.1 | XP_016862785.1 | 6 | 1539 | |||
GenBank mRNA | AK225875.1 | - | 7 | 771 | ||
AK291183.1 | - | 6 | 1028 | |||
AK295508.1 | - | 5 | 1118 | |||
BC004178.2 | - | 6 | 976 | 2959536 | ||
BC004211.2 | - | 6 | 976 | 2959536 | ||
BC019582.1 | - | 6 | 1225 | 3872331 | ||
BC021975.2 | - | 2 | 226 |