Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010000.2 | NP_001010000.1 | 7 | 881 | ||
XM_005258144.1 | XP_005258201.2 | 8 | 1203 | |||
XM_005258145.1 | XP_005258202.2 | 8 | 1203 | |||
XM_005258146.2 | XP_005258203.1 | 8 | 1203 | |||
GenBank mRNA | AB037735.1 | - | 8 | 4154 | ||
AK001174.1 | - | 2 | 210 | |||
AK289791.1 | - | 1 | 141 | |||
AK302026.1 | - | 7 | 790 | |||
AK308259.1 | - | 7 | 829 | |||
BC033668.1 | - | 1 | 100 | |||
BC065274.1 | - | 6 | 642 | 6164396 | ||
BX648684.1 | - | 7 | 839 |