Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136026.2 | NP_001129498.1 | 4 | 426 | ||
NM_001136232.2 | NP_001129704.1 | 3 | 280 | |||
NM_001278946.1 | NP_001265875.1 | 3 | 248 | |||
NM_030673.3 | NP_109598.2 | 4 | 282 | |||
NM_183352.2 | NP_899195.1 | 3 | 248 | |||
XM_005265379.2 | XP_005265436.1 | 4 | 299 | |||
XM_017007019.1 | XP_016862508.1 | 4 | 336 | |||
XM_017007020.1 | XP_016862509.1 | 3 | 143 | |||
XM_017007021.1 | XP_016862510.1 | 3 | 153 | |||
GenBank mRNA | AB209554.1 | - | 2 | 95 | ||
AF052155.1 | - | 3 | 280 | |||
AK223019.1 | - | 3 | 133 | |||
AK294662.1 | - | 3 | 153 | |||
AK301999.1 | - | 4 | 350 | |||
BC002634.2 | - | 4 | 163 | 3605545 | ||
BC091506.1 | - | 3 | 158 | 30408210 | ||
BE621430.1 | - | 3 | 234 | |||
CN344033.1 | - | 3 | 260 | |||
EF036494.1 | - | 3 | 147 | |||
L09260.1 | - | 3 | 125 |