Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030780.4 | NP_110407.2 | 3 | 763 | ||
NR_102338.1 | 4 | 872 | ||||
GenBank mRNA | AF283645.1 | - | 3 | 493 | ||
AK027531.1 | - | 3 | 532 | |||
AK027787.1 | - | 3 | 497 | |||
AK125492.1 | - | 4 | 481 | |||
AL046268.1 | - | 3 | 333 | |||
BC021893.1 | - | 3 | 471 | 4524907 |