Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTDSS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030783.1 | NP_110410.1 | 2, 3 | 470 | ||
XM_005253153.1 | XP_005253210.1 | 2, 3 | 403 | |||
XM_011520391.2 | XP_011518693.1 | 2, 3 | 1040 | |||
XM_011520392.2 | XP_011518694.1 | 3, 4 | 467 | |||
XM_017018368.1 | XP_016873857.1 | 4, 5 | 2862 | |||
XM_017018369.1 | XP_016873858.1 | 3, 4 | 592 | |||
GenBank mRNA | AK311723.1 | - | 2, 3 | 402 | ||
AL834357.1 | - | 2, 3 | 384 | |||
BC001210.1 | - | 2, 3 | 470 | 3503123 |