Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPH6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030785.3 | NP_110412.1 | 4 | 1895 | ||
XM_011527351.1 | XP_011525653.1 | 4 | 1871 | |||
GenBank mRNA | AK093068.1 | - | 3 | 1165 | ||
AK098837.1 | - | 4 | 1840 | |||
AK223347.1 | - | 4 | 1840 | |||
AL136761.1 | - | 4 | 1867 | |||
BC057785.1 | - | 4 | 1723 | |||
BC136351.1 | - | 4 | 1776 | 9020338 | ||
BC136352.1 | - | 4 | 1776 | 9020339 |