Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258392.2 | NP_001245321.1 | 1, 2 | 620 | ||
NM_001258393.2 | NP_001245322.1 | 1, 2 | 620 | |||
NM_001258394.2 | NP_001245323.1 | 1, 2 | 620 | |||
NM_030813.5 | NP_110440.1 | 1, 2 | 620 | |||
XM_005274320.1 | XP_005274377.1 | 1, 2 | 493 | |||
XM_011545289.2 | XP_011543591.1 | 1, 2 | 493 | |||
GenBank mRNA | AK023214.1 | - | 1, 2 | 470 | ||
AK302006.1 | - | 1, 2 | 463 | |||
AK302069.1 | - | 1, 2 | 318 | |||
AL136909.1 | - | 1, 2 | 519 | |||
BC006404.2 | - | 1, 2 | 460 | 4304523 | ||
BI560251.1 | - | 624 | ||||
BU848809.1 | - | 1, 2 | 466 |