Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPEB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288819.1 | NP_001275748.1 | 2 | 631 | ||
NM_030594.4 | NP_085097.3 | 2 | 323 | |||
GenBank mRNA | AF329402.1 | - | 2 | 494 | ||
AK300105.1 | - | 2 | 631 | |||
BC050629.1 | - | 2 | 284 | 6047179 | ||
BX649159.1 | - | 3 | 2063 | |||
HY007452.1 | - | 2 | 324 |