Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXJ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001454.3 | NP_001445.2 | 3 | 2046 | ||
GenBank mRNA | BC046460.1 | - | 3 | 2037 | 5171637 | |
X99349.1 | - | 2 | 1696 |